missing translation for 'onlineSavingsMsg'
Learn More

Tris(3-chlorophenyl)phosphine, 98%

Product Code. 11473514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473514 5 g 5g
11463514 1 g 1g
11483514 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473514 Supplier Thermo Scientific Alfa Aesar Supplier No. A11203.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29949-85-7 | C18H12Cl3P | 365.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29949-85-7
Molecular Formula C18H12Cl3P
Molecular Weight (g/mol) 365.62
MDL Number MFCD00013632
InChI Key QAPGHLJQIVDTPT-UHFFFAOYSA-N
Synonym tris 3-chlorophenyl phosphine, tris 3-chlorophenyl phosphane, tri m-chlorophenyl phosphine, tri-3-chlorophenyl phosphine, acmc-1cg99, phosphine, tris 3-chlorophenyl, tri 3-chlorophnyl phosphine, tri 3-chlorophenyl phosphine
PubChem CID 121600
IUPAC Name tris(3-chlorophenyl)phosphane
SMILES ClC1=CC(=CC=C1)P(C1=CC(Cl)=CC=C1)C1=CC=CC(Cl)=C1

Specifications

Melting Point 64°C to 66°C
Quantity 5 g
Formula Weight 365.63
Percent Purity 98%
Chemical Name or Material Tris (3-chlorophenyl)phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.