missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-thienyl)phosphine, 98+%

Product Code. 11495363
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495363 5 g 5g
11485363 1 g 1g
11405373 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495363 Supplier Thermo Scientific Alfa Aesar Supplier No. A13379.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24171-89-9 | C12H9PS3 | 280.36 g/mol

It employed as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical and organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool, dry place in tightly closed container. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24171-89-9
Molecular Formula C12H9PS3
Molecular Weight (g/mol) 280.36
MDL Number MFCD00015471
InChI Key KUCPTMZJPDVWJL-UHFFFAOYSA-N
Synonym tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine #
PubChem CID 90384
IUPAC Name trithiophen-2-ylphosphane
SMILES S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1

Specifications

Melting Point 28°C to 30°C
Odor Recognizable
Quantity 5 g
Beilstein 203736
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 280.37
Percent Purity ≥98%
Chemical Name or Material Tris(2-thienyl)phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.