missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-phenylpyridine)iridium(III), Thermo Scientific Chemicals

Product Code. 11359818
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359818 0.5 g 0.5g
11369818 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359818 Supplier Thermo Scientific Alfa Aesar Supplier No. 044282.02
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As a green light emitting material

As a green light emitting material

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693794-98-8
Molecular Formula C33H24IrN3
Molecular Weight (g/mol) 654.80
MDL Number MFCD04039766
InChI Key JAZWLEYGTKXZCJ-UHFFFAOYSA-N
Synonym tris 2-phenylpyridine iridium iii
PubChem CID 71433583
IUPAC Name iridium;2-phenylpyridine
SMILES [Ir].C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1

Specifications

Melting Point 326°C to 328°C
Color Yellow
Quantity 0.5 g
Formula Weight 654.82
Physical Form Powder
Chemical Name or Material Tris (2-phenylpyridine)iridium (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.