missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-methyl-4-diethylaminophenyl)methane, 93+%, Thermo Scientific™

Product Code. 10661885 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10661885 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10661885 Supplier Thermo Scientific Acros Supplier No. 375020010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4482-70-6
Melting Point 163°C to 165°C
Assay Percent Range 93% min. (HPLC)
Packaging Glass bottle
Molecular Formula C34H49N3
Linear Formula [(CH3CH2)2NC6H3(CH3)]3CH
Quantity 1 g
Synonym benzenamine, 4,4',4-methylidynetris n,n-diethyl-3-methyl, 4,4',4-methylidynetris n,n-diethyl-m-toluidine, tris 2-methyl-4-diethylaminophenyl methane, tris 4-diethylamino-2-methylphenyl methane, 4,4',4-methanetriyltris n,n-diethyl-3-methylaniline, 4-bis 4-diethylamino-2-methylphenyl methyl-n,n-diethyl-3-methylaniline, 4-bis 4-diethylamino-2-methyl-phenyl methyl-n,n-diethyl-3-methyl-aniline
Solubility Information Solubility in water: insoluble
InChI Key OKJSFKIUVDXFMS-UHFFFAOYSA-N
SMILES CCN(CC)C1=CC(=C(C=C1)C(C2=C(C=C(C=C2)N(CC)CC)C)C3=C(C=C(C=C3)N(CC)CC)C)C
IUPAC Name 4-[bis[4-(diethylamino)-2-methylphenyl]methyl]-N,N-diethyl-3-methylaniline
Molecular Weight (g/mol) 499.78
PubChem CID 78234
Formula Weight 499.78
Percent Purity 93+%
Chemical Name or Material Tris(2-methyl-4-diethylaminophenyl)methane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.