missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-methoxyphenyl)phosphine, 98%

Product Code. 11408973
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408973 5 g 5g
11498963 1 g 1g
11418973 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408973 Supplier Thermo Scientific Alfa Aesar Supplier No. A11096.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4731-65-1 | C21H21O3P | 352.37 g/mol

Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4731-65-1
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00014892
InChI Key IIOSDXGZLBPOHD-UHFFFAOYSA-N
Synonym tris 2-methoxyphenyl phosphine, tris o-methoxyphenyl phosphine, tris 2-methoxyphenyl phosphane, phosphine, tris methoxyphenyl, tris-2-methoxyphenyl-phosphine, tris o-anisyl phosphine, tri-2-methoxyphenyl phosphine, pubchem19707, tri 2-methoxyphenyl phosphine, ksc286k3b
PubChem CID 78464
IUPAC Name tris(2-methoxyphenyl)phosphane
SMILES COC1=CC=CC=C1P(C2=CC=CC=C2OC)C3=CC=CC=C3OC

Specifications

Melting Point 196°C to 204°C
Quantity 5 g
Beilstein 2776186
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 352.37
Percent Purity 98%
Chemical Name or Material Tris (2-methoxyphenyl)phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.