missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-methoxyphenyl)phosphine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 196°C to 204°C |
Quantity | 1 g |
Beilstein | 2776186 |
Sensitivity | Air Sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 352.37 |
Percent Purity | 98% |
Chemical Name or Material | Tris (2-methoxyphenyl)phosphine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'