Learn More
Tris (2-cyanoethyl)phosphine, 95%, Thermo Scientific Chemicals
£21.00 - £331.00
Chemical Identifiers
CAS | 4023-53-4 |
---|---|
Molecular Formula | C9H12N3P |
Molecular Weight (g/mol) | 193.19 |
InChI Key | CHZAMJVESILJGH-UHFFFAOYSA-N |
Synonym | tris 2-cyanoethyl phosphine, 3,3',3-phosphinetriyltripropanenitrile, tris-2-cyanoethyl phosphine, tris-2-cyanoethyl-phosphine, propanenitrile, 3,3',3-phosphinidynetris, acmc-20ajc3, p ch2ch2cn 3, 3-bis 2-cyanoethyl phosphanyl propanenitrile, propanenitrile,3',3-phosphinidynetris, tris 2-cyanoethyl phosphine 10g |
PubChem CID | 77639 |
IUPAC Name | 3-[bis(2-cyanoethyl)phosphanyl]propanenitrile |
SMILES | C(CP(CCC#N)CCC#N)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15254774
|
Thermo Scientific Acros
449710010 |
1 g |
£21.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15264774
|
Thermo Scientific Acros
449710100 |
10 g |
£91.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15274774
|
Thermo Scientific Acros
449710500 |
50 g |
£331.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4023-53-4 | |
193.19 | |
tris 2-cyanoethyl phosphine, 3,3',3-phosphinetriyltripropanenitrile, tris-2-cyanoethyl phosphine, tris-2-cyanoethyl-phosphine, propanenitrile, 3,3',3-phosphinidynetris, acmc-20ajc3, p ch2ch2cn 3, 3-bis 2-cyanoethyl phosphanyl propanenitrile, propanenitrile,3',3-phosphinidynetris, tris 2-cyanoethyl phosphine 10g | |
3-[bis(2-cyanoethyl)phosphanyl]propanenitrile |
C9H12N3P | |
CHZAMJVESILJGH-UHFFFAOYSA-N | |
77639 | |
C(CP(CCC#N)CCC#N)C#N |
Specifications
4023-53-4 | |
235.0°C (0.9 mmHg) | |
Glass bottle | |
tris 2-cyanoethyl phosphine, 3,3',3-phosphinetriyltripropanenitrile, tris-2-cyanoethyl phosphine, tris-2-cyanoethyl-phosphine, propanenitrile, 3,3',3-phosphinidynetris, acmc-20ajc3, p ch2ch2cn 3, 3-bis 2-cyanoethyl phosphanyl propanenitrile, propanenitrile,3',3-phosphinidynetris, tris 2-cyanoethyl phosphine 10g | |
CHZAMJVESILJGH-UHFFFAOYSA-N | |
3-[bis(2-cyanoethyl)phosphanyl]propanenitrile | |
77639 | |
95% |
97.0°C to 99.0°C | |
94% min. (Iodimetry) | |
C9H12N3P | |
Solubility in water: slightly soluble | |
C(CP(CCC#N)CCC#N)C#N | |
193.19 | |
193.19 | |
Tris (2-cyanoethyl)phosphine |
Safety and Handling
GHS H Statement
May cause cancer.
Harmful if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Obtain special instructions before use.
Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 223-687-
- Tris (2-cyanoethyl)phosphine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H350-May cause cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P313-Get medical advice/attention.
RUO – Research Use Only