missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-carboxyethyl)phosphine hydrochloride, 98%

Product Code. 11358558
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358558 0.5 g 0.5g
11378558 2 g 2g
11368558 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358558 Supplier Thermo Scientific Alfa Aesar Supplier No. 040587.02
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reducing agent, specific for disulfides | CAS: 51805-45-9 | C9H12O6P | 247.16 g/mol

Reducing agent, specific for disulfidesTris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides, and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51805-45-9
Molecular Formula C9H12O6P
Molecular Weight (g/mol) 247.16
MDL Number MFCD00145469
InChI Key PZBFGYYEXUXCOF-UHFFFAOYSA-K
Synonym tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
PubChem CID 2734570
SMILES [O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O

Specifications

Melting Point 177°C to 178°C
Color White
Linear Formula (HO2CCH2CH2)3P·HCl
Quantity 0.5 g
UN Number UN3261
Sensitivity Moisture sensitive
Merck Index 14,9085
Solubility Information Soluble in water and methanol.
IUPAC Name 3-[bis(2-carboxylatoethyl)phosphanyl]propanoate
Formula Weight 286.65
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.