missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,3-dibromopropyl) isocyanurate, 97%, Thermo Scientific Chemicals

Product Code. p-7040307
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496518 100 g 100g
15406528 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496518 Supplier Thermo Scientific Alfa Aesar Supplier No. H60056.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tris(2,3-dibromopropyl) isocyanurate is a non degradable heterocyclic hexabrominated flame retardant. It inhibited the expression of psaA and psbC that led to the decrease of Nannochloropsis sp.s growth biomass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2,3-dibromopropyl) isocyanurate is a non degradable heterocyclic hexabrominated flame retardant. It inhibited the expression of psaA and psbC that led to the decrease of Nannochloropsis sp.′s growth biomass.

Solubility
Soluble in chloroform.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52434-90-9
Molecular Formula C12H15Br6N3O3
Molecular Weight (g/mol) 728.694
MDL Number MFCD00010290
InChI Key NZUPFZNVGSWLQC-UHFFFAOYSA-N
Synonym tris 2,3-dibromopropyl isocyanurate, 1,3,5-tris 2,3-dibromopropyl-1,3,5-triazinane-2,4,6-trione, hexahydro-1,3,5-tris 2,3-dibromopropyl-1,3,5-triazine-2,4,6-trione, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3,5-tris 2,3-dibromopropyl, 1,3,5-tris 2,3-dibromopropyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione, isocyanuric acid tris 2,3-dibromopropyl ester, 1,3,5-tris 2,3-dibromopropyl-s-triazine-2,4,6-trione, acmc-1apwt, tri 2,3-dibromopropyl isocyanate
PubChem CID 103634
IUPAC Name 1,3,5-tris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione
SMILES C(C(CBr)Br)N1C(=O)N(C(=O)N(C1=O)CC(CBr)Br)CC(CBr)Br

Specifications

Quantity 100 g
Solubility Information Soluble in chloroform.
Formula Weight 728.69
Percent Purity 97%
Chemical Name or Material Tris(2,3-dibromopropyl) isocyanurate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.