missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, 98+%

Product Code. 11397547
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397547 2 g 2g
11387547 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397547 Supplier Thermo Scientific Alfa Aesar Supplier No. 012783.04
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50525-27-4 | C30H36Cl2N6O6Ru | 748.62 g/mol

Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate is a catalyst used in light-induced redox reactions. It is an important catalyst in studies towards photochemical water splitting. This product can be used as a catalyst in pharmaceutical research. It is widely applied in fields of photoelectrical chemical components, photovoltaic cells, smart biosensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50525-27-4
Molecular Formula C30H36Cl2N6O6Ru
Molecular Weight (g/mol) 748.62
MDL Number MFCD00149670
InChI Key WHELTKFSBJNBMQ-UHFFFAOYSA-L
Synonym tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
PubChem CID 131664161
IUPAC Name 2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Specifications

Melting Point 63°C to 66°C
Color Red to Orange
Assay Percent Range >98%
Quantity 2 g
Solubility Information Soluble in water.
Formula Weight 748.63 (640.54 Anhydrous)
Physical Form Crystalline
Chemical Name or Material Tris(2, 2'-bipyridine)dichlororuthenium(II) hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.