missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)terbium(III), 99%, Thermo Scientific Chemicals

Catalog Number p-7059516
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15492-51-0
C33H57O6Tb
708.74
MFCD00064752
DIIXZXQKWUIOAN-UHFFFAOYSA-N
tb tmhd 3, terbium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate
133109942
[Tb+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
This item is not returnable. View return policy
15492-51-0
C33H57O6Tb
708.74
MFCD00064752
DIIXZXQKWUIOAN-UHFFFAOYSA-N
tb tmhd 3, terbium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate
133109942
[Tb+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 155°C to 156°C
Quantity 1 g
IUPAC Name terbium(3+) tris(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Formula Weight 708.74
Percent Purity 99%
Physical Form Granules
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)terbium (III)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'