missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)scandium(III), 95+%, Thermo Scientific™

Product Code. 11325617
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325617 0.5 g 0.5g
11335617 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325617 Supplier Thermo Scientific Alfa Aesar Supplier No. 042787.02

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Intermediate for scandium oxide ceramics

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15492-49-6
Molecular Formula C33H57O6Sc
Molecular Weight (g/mol) 594.769
MDL Number MFCD00169875
InChI Key SIPNKDOCYGGAQY-LWTKGLMZSA-K
Synonym scandium iii 2,2,6,6-tetramethyl-3,5-heptanedionate, tris 2,2,6,6-tetramethyl-3,5-heptanedionato scandium, 4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy scandio oxy-2,2,6,6-tetramethylhept-4-en-3-one, scandium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate hydrate trace metals basis, tris 2,2,6,6-tetramethyl-3,5-heptanedionato scandium iii-sc reo sc tmhd 3
PubChem CID 134068149
IUPAC Name scandium(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Sc+3]

Specifications

Melting Point 150°C to 152°C
Color White
Boiling Point 275°C (decomposition)
Flash Point >110°C (230°F)
Quantity 0.5 g
Sensitivity Hygroscopic
Solubility Information Soluble in ethyl acetate
Formula Weight 594.77
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)scandium (III)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.