missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), recrystallized, 99.9% (metals basis)

Product Code. 11344837
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344837 25 g 25g
11334837 1 g 1g
11354837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344837 Supplier Thermo Scientific Alfa Aesar Supplier No. 041862.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14876-47-2 | C33H60FeO6 | 608.682 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14876-47-2
Molecular Formula C33H60FeO6
Molecular Weight (g/mol) 608.682
MDL Number MFCD00067465
InChI Key WLITYJBILWOYFF-QFVJJVGWSA-N
Synonym fe tmhd 3
PubChem CID 15251600
IUPAC Name (E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;iron
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Fe]

Specifications

Melting Point 148°C
Color Red
Boiling Point 300°C (decomposition)
Odor Odorless
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 605.66
Percent Purity 99.9%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), Recrystallized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.