missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gadolinium(III)

Product Code. 11362438
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362438 1 g 1g
11352438 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362438 Supplier Thermo Scientific Alfa Aesar Supplier No. 014486.03
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14768-15-1 | C33H57GdO6 | 707.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14768-15-1
Molecular Formula C33H57GdO6
Molecular Weight (g/mol) 707.06
MDL Number MFCD00010349
InChI Key UIVVGOMXFUOLAN-UHFFFAOYSA-N
Synonym tris 2,2,6,6-tetramethyl-3,5-heptanedionate gadolinium
PubChem CID 132984032
IUPAC Name gadolinium;(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
SMILES [Gd+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Specifications

Melting Point 167°C to 170°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 707.07
Physical Form Crystalline
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gadolinium(III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.