missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III), 98%, Thermo Scientific™

Product Code. 11397918
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397918 0.5 g 0.5g
11307928 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397918 Supplier Thermo Scientific Alfa Aesar Supplier No. 036464.02
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Superconductor MOCVD Precursor

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15522-69-7
Molecular Formula C33H60DyO6
Molecular Weight (g/mol) 715.337
MDL Number MFCD00000017
InChI Key RBHZQBMBFRBKKB-LWTKGLMZSA-N
Synonym Dysprosium(III)-DPM; Dy(TMHD)3
PubChem CID 72376403
IUPAC Name dysprosium;(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Dy]

Specifications

Melting Point 182°C to 183°C
Quantity 0.5 g
Sensitivity Hygroscopic
Formula Weight 712.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.