missing translation for 'onlineSavingsMsg'
Learn More

Tris(1-methoxy-2-methyl-2-propoxy)bismuth, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15426965
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tris(1-methoxy-2-methyl-2-propoxy)bismuth is used as metal organic precursors to prepare bismuth titanium oxide thin films, which finds application in ferroelectric random access memory.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H33O6Bi
MDL Number MFCD11113030

Specifications

Melting Point 70°C
Color Yellow
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in ethylcyclohexane.
Formula Weight 518.4
Percent Purity 99.99%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Tris (1-methoxy-2-methyl-2-propoxy)bismuth

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'