missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(1-chloro-2-propyl) Phosphate-d18, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Tris(1-chloro-2-propyl) Phosphate-d18 |
| CAS | 1447569-78-9 |
| Molecular Formula | C9D18Cl3O4P |
| Quantity | 10 mg |
| Synonym | 1-Chloro-2-propanol-d6 2,2',2''-phosphate, TCPP-d18, Tris(1-chloro-2-propyl-1,2,2,3,3,3-d6) phosphate, Tris[2-chloro-1-(methyl-d3)ethyl-1,2,2-d3] phosphate, Tris(2-chloroisopropyl) phosphate-d18, Tris(2-chloroisopropyl) phosphate D18, Tris(chloroisopropyl) phosphate-d18, Tris(β-chloroisopropyl) phosphate-d18, Tris(1-chloro-2-propyl) phosphate-d18 |
| InChI Formula | InChI=1S/C9H18Cl3O4P/c1-7(4-10)14-17(13,15-8(2)5-11)16-9(3)6-12/h7-9H,4-6H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D,8D,9D |
| SMILES | [2H]C([2H])([2H])C([2H])(OP(=O)(OC([2H])(C([2H])([2H])[2H])C([2H])([2H])Cl)OC([2H])(C([2H])([2H])[2H])C([2H])([2H])Cl)C([2H])([2H])Cl |
| Molecular Weight (g/mol) | 345.68 |
| Formula Weight | 344.1138 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction