missing translation for 'onlineSavingsMsg'
Learn More

Tris(1,3-dichloro-2-propyl) Phosphate-d15, TRC

Product Code. 30452728
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452728 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30452728 Supplier Toronto Research Chemicals Supplier No. TRCT87630725MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris(1,3-dichloro-2-propyl) phosphate-d15
CAS 1447569-77-8
Purity Grade Notes HPLC
Molecular Formula C9 D15 Cl6 O4 P
Quantity 25 mg
Synonym Tris(2-chloro-1-(chloromethyl)ethyl) phosphate D15, Tris(1,3-dichloro-2-propyl) phosphate D15, Tris(1,3-dichloroisopropyl) phosphate D15
InChI Formula InChI=1S/C9H15Cl6O4P/c10-1-7(2-11)17-20(16,18-8(3-12)4-13)19-9(5-14)6-15/h7-9H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D,8D,9D
SMILES [2H]C([2H])(Cl)C([2H])(OP(=O)(OC([2H])(C([2H])([2H])Cl)C([2H])([2H])Cl)OC([2H])(C([2H])([2H])Cl)C([2H])([2H])Cl)C([2H])([2H])Cl
IUPAC Name tris[2-chloro-1-[chloro(dideuterio)methyl]-1,2,2-trideuterio-ethyl] phosphate
Molecular Weight (g/mol) 446
Formula Weight 442.978
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.