missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TRIS, 1.0M buffer soln., pH 7.6
Click to view available options
Quantity:
1 L
Description
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
Store in cool place. Keep container
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
Store in cool place. Keep container
Specifications
CAS | 77-86-1 |
Concentration | 1.0 M |
pH | 7.6 |
Physical Form | Liquid |
Quantity | 1 L |
Molecular Formula | C4H11NO3 |
Odor | Odorless |
MDL Number | MFCD00004679 |
Solubility Information | Soluble in water and organic solvents. |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'