missing translation for 'onlineSavingsMsg'
Learn More

Triricinolein, TRC

Product Code. 30138339
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138339 50 mg 50mg
30135000 100 mg 100mg
30005619 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138339 Supplier Toronto Research Chemicals Supplier No. TRCT90205050MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2540-54-7
Molecular Formula C57H104O9
Molecular Weight (g/mol) 933.43
Synonym (9Z,9'Z,9' 'Z,12R,12'R,12' 'R)- (9CI)-12-Hydroxy-1,2,3-propanetriyl Ester 9-Octadecenoic Acid, 12-Hydroxy-1,2,3-propanetriyl Ester 9-Octadecenoic Acid Stereoisomer, (6CI,7CI,8CI)-Tri-Ricinolein, 1,2,3-Tris[((9Z,12R)-12-hydroxy-9-octadecenoyl)oxy]propane, Geopon GO 13, Glycerin triricinoleate, Glycerol triricinoleate, Glyceryl triricinoleate, NSC 93749, Ricinoleic acid triglyceride, Ricinoleic triglyceride
IUPAC Name 2,3-bis[[(Z,12R)-12-hydroxyoctadec-9-enoyl]oxy]propyl (Z,12R)-12-hydroxyoctadec-9-enoate
SMILES CCCCCC[C@@H](O)C\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/C[C@H](O)CCCCCC)OC(=O)CCCCCCC\C=C/C[C@H](O)CCCCCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C57H104O9/c1-4-7-10-31-40-51(58)43-34-25-19-13-16-22-28-37-46-55(61)64-49-54(66-57(63)48-39-30-24-18-15-21-27-36-45-53(60)42-33-12-9-6-3)50-65-56(62)47-38-29-23-17-14-20-26-35-44-52(59)41-32-11-8-5-2/h25-27,34-36,51-54,58-60H,4-24,28-33,37-50H2,1-3H3/b34-25-,35-26-,36-27-/t51-,52-,53-/m1/s1
Formula Weight 932.768
Chemical Name or Material Triricinolein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.