missing translation for 'onlineSavingsMsg'
Learn More

Tripropylene glycol

Product Code. 11358287
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
Unit Size:
1 L
250mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358287 250 mL 250mL
11368287 1 L 1 L
11378287 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358287 Supplier Thermo Scientific Alfa Aesar Supplier No. 031798.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24800-44-0 | HOCH(CH3)CH2OCH2CH(CH3)OCH2CH(CH3)OH

Tripropylene glycol is used for textile soaps, lubricants, cutting oils and greases. It acts as an intermediate in the preparation of urethanes. It is also used as a solvent in coatings and inks. It is utilized as varnish removers, emulsifiers and hydraulic fluids. As an additive, it is used for carburetor fuels. It is involved as a cooling media as well as an active ingredient of plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24800-44-0
Molecular Formula HOCH(CH3)CH2OCH2CH(CH3)OCH2CH(CH3)OH
MDL Number MFCD00014405

Specifications

Density 1.018 g/mL
Boiling Point 268°C
Odor Odorless
Refractive Index 1.4443
Quantity 250 mL
Solubility Information Miscible with water,methanol and ether.
Formula Weight 192.25
Physical Form Liquid
Chemical Name or Material Tripropylene glycol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.