missing translation for 'onlineSavingsMsg'
Learn More

Tripropylene glycol, 99%, pure, mixture of isomers

Product Code. 15506695 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
25 L
Unit Size:
1 L
25 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15506695 1 L 1 L
15525286 5 L 5 L
15535286 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15506695 Supplier Thermo Scientific Acros Supplier No. 458480010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24800-44-0 | C9H20O4 | 192.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24800-44-0
Molecular Formula C9H20O4
Molecular Weight (g/mol) 192.25

Specifications

Density 1.0200g/mL
Boiling Point 270.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (sum of isomers) (GC)
Quantity 1 L
Specific Gravity 1.02
Solubility Information Solubility in water: soluble.
Viscosity 77.3 mm²/s (20°C)
Formula Weight 192.25
Percent Purity 99%
Grade Pure
Chemical Name or Material Tripropylene glycol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.