missing translation for 'onlineSavingsMsg'
Learn More

Tripropylamine, 98%

Product Code. 10216483 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216483 2.5 L Glass bottle 2.5 L
10428720 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216483 Supplier Thermo Scientific Acros Supplier No. 162690025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-69-2 | C9H21N | 143.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-69-2
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD00009363
InChI Key YFTHZRPMJXBUME-UHFFFAOYSA-N
Synonym tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
PubChem CID 7616
ChEBI CHEBI:38880
IUPAC Name N,N-dipropylpropan-1-amine
SMILES CCCN(CCC)CCC

Specifications

Melting Point -93.0°C
Color Colorless
Density 0.7600g/mL
Boiling Point 156.0°C
Flash Point 32.5°C
Infrared Spectrum Authentic
Assay Percent Range 0.2% max. dipropylamine (GC)
Packaging Glass bottle
Linear Formula (CH3CH2CH2)3N
Refractive Index 1.4155 to 1.4175
Quantity 2.5 L
Beilstein 04, II, 623
Specific Gravity 0.76
Solubility Information Solubility in water: 2.6g/L (20°C). Other solubilities: soluble in most organic solvents
Viscosity 0.627 mPa.s (25°C)
Formula Weight 143.27
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tripropylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.