missing translation for 'onlineSavingsMsg'
Learn More

Juchheim Laborgeraete™ Tripod Chain Clamp

Tripod chain clamp with a span of 70 mm.

Brand:  Juchheim Laborgeraete™ 05160

Additional Details : Weight : 0.50000kg

Product Code. 10347140

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Variable applicable
  • For vessel diameters from 10 to 70 mm
  • Particularly durable materials (brass and malleable iron)
Specifications

Specifications

Tempered Cast Iron
70 mm
Span of 70 mm, Chain- 350 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers