Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringe
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
Brand:  Thermo Scientific™ 365H2161
Code : 16
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 12876036
  • £114.26 / Each
In Stock
to see stock.
Description
Description
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
Specifications
Specifications
Stainless Steel (Needle), Stainless Steel (Plunger)
26s G
100 μL
57 mm
TriPlus RSH Autosampler and TriPlus 100LS Autosampler
LV Splitless
Gas Tight GC Autosampler Syringe, Fixed Needle
TriPlus
Cone
Fixed