missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltin chloride 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30326137
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326137 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326137 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010359ME
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fentin-chloride (Triphenyltin chloride)
CAS 639-58-7
Analyte or Component Names Fentin-chloride (Triphenyltin chloride)
Shipping Condition Room Temperature
Molecular Formula C18 H15 Cl Sn
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Stannane, chlorotriphenyl-, Triphenyltin chloride (6CI), Chlorotriphenylstannane, Aquatin, Brestanol, Chlorotriphenyltin, Fentin chloride, Fentin-chloride, GC 8993, General Chemicals 8993, HOE 2872, LS 4442, NSC 1214, NSC 43675, TPTC, Triphenylchlorostannane, Triphenylchlorotin, Triphenylstannyl chloride, Triphenyltin monochloride, Wenqukexing
InChI Formula InChI=1S/3C6H5.ClH.Sn/c3*1-2-4-6-5-3-1;;/h3*1-5H;1H;/q;;;;+1/p-1
Solution Type Single Solution
SMILES Cl[Sn](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name chloro(triphenyl)stannane
Solvent or Matrix Methanol
Industry Type Environmental Testing
Molecular Weight (g/mol) 385.47
Quantity 1 mL
Formula Weight 385.9884 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.