missing translation for 'onlineSavingsMsg'
Learn More

(Triphenylphosphoranylidene)acetonitrile, 96%

Product Code. 10795863 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10795863 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10795863 Supplier Thermo Scientific Acros Supplier No. 371570050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16640-68-9 | C20H16NP | 301.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16640-68-9
Melting Point 189.0°C to 198.0°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Molecular Formula C20H16NP
Linear Formula (C6H5)3P=CHCN
MDL Number MFCD00567633
Quantity 5 g
Synonym triphenylphosphoranylidene acetonitrile, 2-triphenylphosphoranylidene acetonitrile, acetonitrile, triphenylphosphoranylidene, 2-(triphenyl-λ5-phosphanylidene)acetonitrile, 2-triphenyl-lambda5-phosphanylidene acetonitrile, acmc-1c6hd, ksc493a4b, cyanomethylenetriphenylphosphorane, cyanomethylene triphenylphosphorane, triphenylphosphoranylidene acetonitrile, 2-triphenylphosphoranylidene acetonitrile, acetonitrile, triphenylphosphoranylidene, 2-(triphenyl-λ5-phosphanylidene)acetonitrile, 2-triphenyl-lambda5-phosphanylidene acetonitrile, acmc-1c6hd, ksc493a4b, cyanomethylenetriphenylphosphorane, cyanomethylene triphenylphosphorane
InChI Key APISVOVOJVZIBA-UHFFFAOYSA-N
SMILES N#CC=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 2-(triphenyl-λ⁵-phosphanylidene)acetonitrile
Molecular Weight (g/mol) 301.33
PubChem CID 85524
Formula Weight 301.33
Percent Purity 96%
Chemical Name or Material (Triphenylphosphoranylidene)acetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.