missing translation for 'onlineSavingsMsg'
Learn More

(Triphenylphosphoranylidene)acetaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 10237610
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2136-75-6
C20H17OP
304.33
MFCD00006994
CQCAYWAIRTVXIY-UHFFFAOYSA-N
2-triphenylphosphoranylidene acetaldehyde, formylmethylene triphenylphosphorane, triphenylphosphoranylidene acetaldehyde, formylmethylenetriphenylphosphorane, acetaldehyde, triphenylphosphoranylidene, 2-triphenyl-??-phosphanylidene acetaldehyde, acmc-209fjg, ksc492c5l
75051
2-(triphenyl-$l^{5}-phosphanylidene)acetaldehyde
C1=CC=C(C=C1)P(=CC=O)(C2=CC=CC=C2)C3=CC=CC=C3
This item is not returnable. View return policy
2136-75-6
C20H17OP
304.33
MFCD00006994
CQCAYWAIRTVXIY-UHFFFAOYSA-N
2-triphenylphosphoranylidene acetaldehyde, formylmethylene triphenylphosphorane, triphenylphosphoranylidene acetaldehyde, formylmethylenetriphenylphosphorane, acetaldehyde, triphenylphosphoranylidene, 2-triphenyl-??-phosphanylidene acetaldehyde, acmc-209fjg, ksc492c5l
75051
2-(triphenyl-$l^{5}-phosphanylidene)acetaldehyde
C1=CC=C(C=C1)P(=CC=O)(C2=CC=CC=C2)C3=CC=CC=C3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 185.0°C to 188.0°C
Color Brown to Pink
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)3P=CHCHO
Quantity 5 g
Formula Weight 304.33
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material (Triphenylphosphoranylidene)acetaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'