missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine Sulfide, 99+%

Product Code. 10373522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10373522 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10373522 Supplier Thermo Scientific Acros Supplier No. 159030100
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3878-45-3 | C18H15PS | 294.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3878-45-3
Melting Point 162.0°C to 164.0°C
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C18H15PS
Linear Formula (C6H5)3P(S)
MDL Number MFCD00004917
Quantity 10 g
Beilstein 16, 784
Fieser 05,732
Synonym triphenylphosphine sulfide, phosphine sulfide, triphenyl, triphenylphosphine sulphide, triphenylphosphine monosulfide, triphenylphosphane sulfide, triphenylphosphino-1-thione, triphenylthioxophosphorane, triphenylphosphorus sulfide, wln: spr&r&r, triphenyl-, e5-phosphanethione
Solubility Information Solubility in water: practically insoluble. Other solubilities: slightly soluble in organic solvents
InChI Key VYNGFCUGSYEOOZ-UHFFFAOYSA-N
SMILES S=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name triphenyl(sulfanylidene)-$l^{5}-phosphane
Molecular Weight (g/mol) 294.35
PubChem CID 19758
Formula Weight 294.34
Percent Purity 99+%
Chemical Name or Material Triphenylphosphine sulfide, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Triphenylphosphine sulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.