missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine sulfide, 98%, Thermo Scientific Chemicals

Product Code. 11432918
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432918 100 g 100g
11422918 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432918 Supplier Thermo Scientific Alfa Aesar Supplier No. A16884.22
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3878-45-3 | Molecular Formula: C18H15PS | Molecular Weight: 294.35 g/mol

Triphenylphosphine sulfide involves in the , conversion of epoxides to episulfides (thiiranes) in presence of TFA of the same stereochemistry. It acts as a ligand for the Pd-catalyzed bisalkoxycarbonylation of olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylphosphine sulfide involves in the , conversion of epoxides to episulfides (thiiranes) in presence of TFA of the same stereochemistry. It acts as a ligand for the Pd-catalyzed bisalkoxycarbonylation of olefins.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 3878-45-3
Molecular Formula C18H15PS
Molecular Weight (g/mol) 294.35
MDL Number MFCD00004917
InChI Key VYNGFCUGSYEOOZ-UHFFFAOYSA-N
Synonym triphenylphosphine sulfide, phosphine sulfide, triphenyl, triphenylphosphine sulphide, triphenylphosphine monosulfide, triphenylphosphane sulfide, triphenylphosphino-1-thione, triphenylthioxophosphorane, triphenylphosphorus sulfide, wln: spr&r&r, triphenyl-, e5-phosphanethione
PubChem CID 19758
IUPAC Name triphenyl(sulfanylidene)-$l^{5}-phosphane
SMILES S=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifica

Melting Point 158°C to 164°C
Quantity 100 g
UN Number UN3464
Beilstein 645872
Solubility Information Insoluble in water.
Formula Weight 294.36
Percent Purity 98%
Chemical Name or Material Triphenylphosphine sulfide

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.