missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylphosphine, polymer supported 1.4-2.0mmol/g on polystyrene 200-400 mesh 2% DVB loading, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Triphenylphosphine polymer |
| CAS | 39319-11-4 |
| Molecular Formula | (C20H17P.C10H10.C8H8)x |
| Quantity | 1 g |
| Synonym | Triphenylphosphine','Polymer-Bound |
| SMILES | C=CC1=CC=C(P(C2=CC=CC=C2)C3=CC=CC=C3)C=C1.C=CC4=CC=CC=C4.[D1]C=C.[D1]C=C.C5=CC=CC=C5 |
| Molecular Weight (g/mol) | 262.29 |
| Concentration or Composition (by Analyte or Components) | 1.4-2.0 mmol/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction