missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine Oxide, TRC

Product Code. 30452974
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452974 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452974 Supplier Toronto Research Chemicals Supplier No. TRCT808980500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triphenylphosphine oxide
CAS 791-28-6
Purity Grade Notes HPLC
Molecular Formula C18 H15 O P
Quantity 500 mg
Synonym Triphenylphosphine oxide (ACI), (Diphenylphosphoroso)benzene, NSC 398, PP 560, TPPO, Triphenyl phosphorus oxide, Triphenylphosphine monoxide, Phosphine oxide','triphenyl- (6CI','8CI','9CI','ACI)
InChI Formula InChI=1S/C18H15OP/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H
SMILES O=P(c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name diphenylphosphorylbenzene
Molecular Weight (g/mol) 278.28
Formula Weight 278.0861
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.