missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine oxide, 99%

Product Code. 11469683
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11469683

Brand: Thermo Scientific Alfa Aesar A12455.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 791-28-6 | C18H15OP | 278.29 g/mol

Triphenylphosphine oxide is used as a catalyst, Lewis base, ligand for metals and starting material for the preparation of other phosphorus compounds. It is also used in epoxidations, Michael reactions and coupling reactions. It reacts with copper(II) halides and form coordination complexes of Copper(II). Further, it acts as a crystallization aid for chemical compounds. In addition to this, it is used for processing, extraction, separation and spectrophotometric determination of cadmium and mercury.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylphosphine oxide is used as a catalyst, Lewis base, ligand for metals and starting material for the preparation of other phosphorus compounds. It is also used in epoxidations, Michael reactions and coupling reactions. It reacts with copper(II) halides and form coordination complexes of Copper(II). Further, it acts as a crystallization aid for chemical compounds. In addition to this, it is used for processing, extraction, separation and spectrophotometric determination of cadmium and mercury.

Solubility
Soluble in polar organic solvents. Slightly soluble in hexane and diethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 791-28-6
Molecular Formula C18H15OP
Molecular Weight (g/mol) 278.29
MDL Number MFCD00002080 MFCD03458802
InChI Key FIQMHBFVRAXMOP-UHFFFAOYSA-N
Synonym triphenylphosphine oxide, phosphine oxide, triphenyl, triphenyl phosphorus oxide, triphenyl phosphine oxide, triphenylphosphane oxide, tppo, triphenylphosphineoxide, ph3po, diphenylphosphoroso benzene, triphenylphosphanoxid
PubChem CID 13097
ChEBI CHEBI:36601
IUPAC Name diphenylphosphorylbenzene
SMILES O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 153°C to 159°C
Density 1.2
Boiling Point 360°C
Flash Point 180°C (356°F)
Quantity 100 g
Beilstein 745854
Solubility Information Soluble in polar organic solvents. Slightly soluble in hexane and diethyl ether. Insoluble in water.
Formula Weight 278.29
Percent Purity 99%
Chemical Name or Material Triphenylphosphine oxide
compliance-icons
Product Identifier
  • Triphenylphosphine oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Triphenylphosphine oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.