missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine hydrobromide, 97%

Product Code. 11389085
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389085 5 g 5g
11399085 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389085 Supplier Thermo Scientific Alfa Aesar Supplier No. L14290.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6399-81-1 | C18H16BrP | 343.20 g/mol

Triphenylphosphine hydrobromide is used as mild source of anhydrous HBr; catalyst for formation of THP ethers from tertiary alcohols; preparation of phosphonium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylphosphine hydrobromide is used as mild source of anhydrous HBr; catalyst for formation of THP ethers from tertiary alcohols; preparation of phosphonium salts.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6399-81-1
Molecular Formula C18H16BrP
Molecular Weight (g/mol) 343.20
MDL Number MFCD00035107
InChI Key CMSYDJVRTHCWFP-UHFFFAOYSA-N
Synonym triphenylphosphine hydrobromide, triphenylphosphonium bromide, phosphine, triphenyl-, hydrobromide, triphenylphosphane hydrobromide, triphenylphosphine bromine, pubchem13868, ph3p hbr, ph3p . hbr, unappfzuzwmopv-uhfffaoysa-n
PubChem CID 80811
IUPAC Name triphenylphosphane;hydrobromide
SMILES Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼196°C
Odor Odorless
Quantity 5 g
UN Number UN3261
Beilstein 3633387
Formula Weight 343.21
Percent Purity 97%
Chemical Name or Material Triphenylphosphine hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.