missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine dibromide, ca. 33% bromine

Product Code. 10134830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10134830 25 g Plastic bottle 25g
10787583 5 g Plastic bottle 5g
10548883 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10134830 Supplier Thermo Scientific Acros Supplier No. 291070250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1034-39-5 | C18H15Br2P | 422.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1034-39-5
Molecular Formula C18H15Br2P
Molecular Weight (g/mol) 422.10
MDL Number MFCD00000054
InChI Key OCXGTPDKNBIOTF-UHFFFAOYSA-N
Synonym dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane
PubChem CID 70579
IUPAC Name dibromo(triphenyl)-$l^{5}-phosphane
SMILES BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 235.0°C
Color Pink to Yellow
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula (C6H5)3PBr2
Quantity 25 g
Beilstein 16, 864
Fieser 01,1247; 13,333
Merck Index 15, 9917
Formula Weight 422.09
Percent Purity ca. 33% bromine
Physical Form Crystalline Powder
Chemical Name or Material Triphenylphosphine dibromide, ca. 33% bromine
Show More Show Less
compliance-icons
Product Identifier
  • Triphenylphosphine dibromide, ca. 33% bromine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.