missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine-d15, TRC

Product Code. 30454742
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454742 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30454742 Supplier Toronto Research Chemicals Supplier No. TRCT80897750MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Triphenylphosphine-d15
CAS 24762-44-5
Molecular Formula C18 D15 P
Quantity 50 mg
Synonym tris(2,3,4,5,6-Pentadeuteriophenyl)phosphane-d15, Phosphine','tri(phenyl-2,3,4,5,6-d5)- (ACI), Phosphine','tri(phenyl-d5)- (8CI','9CI), Tri(phenyl-2,3,4,5,6-d5)phosphine (ACI), Perdeuterotriphenylphosphine, Tris(pentadeuterophenyl)phosphine
InChI Formula InChI=1S/C18H15P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D,15D
SMILES [2H]c1c([2H])c([2H])c(P(c2c([2H])c([2H])c([2H])c([2H])c2[2H])c3c([2H])c([2H])c([2H])c([2H])c3[2H])c([2H])c1[2H]
IUPAC Name tris(2,3,4,5,6-pentadeuteriophenyl)phosphane
Molecular Weight (g/mol) 277.38
Formula Weight 277.1853
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.