missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine-3,3',3″-trisulfonic acid trisodium salt hydrate, tech. 85%

Product Code. 11410231
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410231 0.25 g 0.25g
11308338 1 g 1g
11398328 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410231 Supplier Thermo Scientific Alfa Aesar Supplier No. 039538.01
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

In the purification of precious metals | CAS: 335421-90-4 | C18H12Na3O9PS3 | 568.41 g/mol

Triphenylphosphine-3,3',3″-trisulfonic acid trisodium salt hydrate is useful for the preparation of water-soluble organometal catalysts, which finds application in the hydroformylation of propene. It acts as a Lewis base, which is stronger than triphenylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335421-90-4
Molecular Formula C18H12Na3O9PS3
Molecular Weight (g/mol) 568.41
MDL Number MFCD00145472
InChI Key MYAJTCUQMQREFZ-UHFFFAOYSA-K
Synonym sodium 3,3',3-phosphinetriyltribenzenesulfonate, tppts, triphenylphosphine-3,3',3-trisulfonic acid trisodium salt, tris 3-sulfophenyl phosphine trisodium salt, trisodium 3,3',3-phosphinetriyltris benzene-1-sulphonate, trisodium 3-bis 3-sulfonatophenyl phosphanylbenzenesulfonate, tri m-sulfophenyl phosphine, sodium salt, benzenesulfonic acid, 3,3',3-phosphinidynetris-, trisodium salt, triphenylphosphine-3,3 ,3-trisulfonic acid trisodium salt, pubchem16013
PubChem CID 6099338
IUPAC Name trisodium;3-bis(3-sulfonatophenyl)phosphanylbenzenesulfonate

Specifications

Quantity 0.25 g
Beilstein 3584807
Solubility Information Soluble in water.
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)P(C1=CC(=CC=C1)S([O-])(=O)=O)C1=CC(=CC=C1)S([O-])(=O)=O
Formula Weight 568.41 (Anhydrous)
Percent Purity 85%
Grade Technical
Physical Form Crystalline
Chemical Name or Material Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.