missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethylamine, TRC

Product Code. 30455733
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455733 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455733 Supplier Toronto Research Chemicals Supplier No. TRCT808960250G
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triphenylmethylamine
CAS 5824-40-8
Molecular Formula C19H17N
Quantity 250 g
Synonym α,α-Diphenylbenzenemethanamine, Tritylamine, Aminotriphenylmethane, NSC 1154, Triphenylmethanamine
InChI Formula InChI=1S/C19H17N/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H,20H2
SMILES NC(c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name triphenylmethanamine
Molecular Weight (g/mol) 259.35
Formula Weight 259.1361
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.