missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethyl chloride, 98%

Product Code. 10185723 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10185723

Brand: Thermo Scientific Acros 140400025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76-83-5 | C19H15Cl | 278.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-83-5
Molecular Formula C19H15Cl
Molecular Weight (g/mol) 278.78
MDL Number MFCD00000813,MFCD00284810
InChI Key JBWKIWSBJXDJDT-UHFFFAOYSA-N
Synonym trityl chloride, triphenylmethyl chloride, triphenylchloromethane, chlorotriphenylmethane, chloromethanetriyl tribenzene, triphenylmethylchloride, methane, chlorotriphenyl, triphenyl chloromethane, chlorodiphenylmethyl benzene, benzene, 1,1',1-chloromethylidyne tris
PubChem CID 6456
SMILES ClC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 110.0°C to 113.0°C
Color Beige-Yellow or White
Boiling Point 230.0°C to 235.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5)3CCl
Quantity 2.5 kg
Beilstein 05, 700
Fieser 01,1254; 02,453; 04,565; 05,741; 06,656; 15,359
Merck Index 15, 9944
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene, dichloromethane and dmf
IUPAC Name (chlorodiphenylmethyl)benzene
Formula Weight 278.77
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Triphenylmethyl chloride, 98%
Show More Show Less
compliance-icons
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.