missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethanol, Mikromol™

Product Code. 17900476
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
17900476 25 mg 25mg
17933616 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 17900476

Brand: Mikromol MM0173.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Triphenylmethanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triphenylmethanol
CAS 76-84-6
Analyte or Component Names Triphenylmethanol
Shipping Condition Room Temperature
Molecular Formula C19 H16 O
Certifications/Compliance ISO 17025
Synonym Losartan Potassium Imp. G (EP), Losartan Imp. G (EP), Zidovudine Imp. D (EP), Triphenylmethanol, Triphenylmethanol, Ph Eur Zidovudine Impurity D, Losartan Potassium Impurity G, Zidovudine Impurity D, Losartan Impurity G
InChI Formula InChI=1S/C19H16O/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,20H
SMILES OC(c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name triphenylmethanol
Molecular Weight (g/mol) 260.33
Quantity 25 mg
Formula Weight 260.1201 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.