missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethanol, 98%, Thermo Scientific Chemicals

Catalog Number 11444173
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
76-84-6
C19H16O
260.34
MFCD00004445,MFCD10565638
LZTRCELOJRDYMQ-UHFFFAOYSA-N
triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl
6457
triphenylmethanol
OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
76-84-6
C19H16O
260.34
MFCD00004445,MFCD10565638
LZTRCELOJRDYMQ-UHFFFAOYSA-N
triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl
6457
triphenylmethanol
OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.

Solubility
Soluble in dioxane, ethanol, water, ether and benzene.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 161°C to 164°C
Boiling Point 360°C to 380°C
Assay Percent Range 98%
Quantity 50 g
Beilstein 1460837
Merck Index 14,9739
Solubility Information Soluble in dioxane,ethanol,water,ether and benzene.
Formula Weight 260.34
Percent Purity 98%
Chemical Name or Material Triphenylmethanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'