missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethanol, 98%

Product Code. 11434173
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434173 1000 g 1000g
11444173 50 g 50g
11454173 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434173 Supplier Thermo Scientific Alfa Aesar Supplier No. A10366.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76-84-6 | C19H16O | 260.34 g/mol

Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.

Solubility
Soluble in dioxane, ethanol, water, ether and benzene.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-84-6
Molecular Formula C19H16O
Molecular Weight (g/mol) 260.34
MDL Number MFCD00004445,MFCD10565638
InChI Key LZTRCELOJRDYMQ-UHFFFAOYSA-N
Synonym triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl
PubChem CID 6457
IUPAC Name triphenylmethanol
SMILES OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 161°C to 164°C
Boiling Point 360°C to 380°C
Assay Percent Range 98%
Quantity 1000 g
Beilstein 1460837
Merck Index 14,9739
Solubility Information Soluble in dioxane,ethanol,water,ether and benzene.
Formula Weight 260.34
Percent Purity 98%
Chemical Name or Material Triphenylmethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.