missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethane, 99+%

Product Code. 10346500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346500 100 g Glass bottle 100g
10041670 25 g Glass bottle 25g
13468368 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10346500 Supplier Thermo Scientific Acros Supplier No. 146381000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 519-73-3 | C19H16 | 244.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 519-73-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00004763
InChI Key AAAQKTZKLRYKHR-UHFFFAOYSA-N
Synonym triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
PubChem CID 10614
ChEBI CHEBI:76212
IUPAC Name benzhydrylbenzene
SMILES C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 90.0°C to 94.0°C
Color Yellow
Density 1.0100g/mL
Boiling Point 358.0°C to 359.0°C (754.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)3CH
Quantity 100 g
Beilstein 05,698
Merck Index 15,9914
Specific Gravity 1.01
Solubility Information Solubility in water: insoluble in water
Formula Weight 244.34
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Triphenylmethane, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.