missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethane, 98%

Product Code. 11439223
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439223 100 g 100g
11429223 25 g 25g
11449223 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439223 Supplier Thermo Scientific Alfa Aesar Supplier No. A11710.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 519-73-3 | C19H16 | 244.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 519-73-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00004763
InChI Key AAAQKTZKLRYKHR-UHFFFAOYSA-N
Synonym triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
PubChem CID 10614
ChEBI CHEBI:76212
IUPAC Name benzhydrylbenzene
SMILES C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 92°C to 94°C
Density 1.01
Boiling Point 358°C to 359°C
Quantity 100 g
Beilstein 1909753
Merck Index 14,9741
Formula Weight 244.34
Percent Purity 98%
Chemical Name or Material Triphenylmethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.