missing translation for 'onlineSavingsMsg'
Learn More

Triphenylene, 98%

Product Code. 11334384
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334384 1 g 1g
11344384 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334384 Supplier Thermo Scientific Alfa Aesar Supplier No. L00955.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 217-59-4 | C18H12 | 228.29 g/mol

Triphenylene is used in optics and electronics. It is also used as a discotic mesogen in liquid crystalline materials. Further, it is a compound that fluoresces in the ultraviolet region. In addition to this, it is used in the preparation of triphenylene-2-carbaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylene is used in optics and electronics. It is also used as a discotic mesogen in liquid crystalline materials. Further, it is a compound that fluoresces in the ultraviolet region. In addition to this, it is used in the preparation of triphenylene-2-carbaldehyde.

Solubility
Soluble in ethanol, benzene, acetic acid, oils and chloroform.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 217-59-4
Molecular Formula C18H12
Molecular Weight (g/mol) 228.29
MDL Number MFCD00001108
InChI Key SLGBZMMZGDRARJ-UHFFFAOYSA-N
Synonym 9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl
PubChem CID 9170
ChEBI CHEBI:33080
IUPAC Name triphenylene
SMILES C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21

Specifications

Melting Point 194°C to 199°C
Density 1.302
Boiling Point 438°C
Quantity 1 g
Beilstein 1342908
Merck Index 14,9740
Solubility Information Soluble in ethanol,benzene,acetic acid,oils and chloroform.
Formula Weight 228.3
Percent Purity 98%
Chemical Name or Material Triphenylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.