missing translation for 'onlineSavingsMsg'
Learn More

Triphenylcarbenium tetrafluoroborate, 97%

Product Code. 11479793
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479793 250 g 250g
11459793 10 g 10g
11469793 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479793 Supplier Thermo Scientific Alfa Aesar Supplier No. A12949.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 341-02-6 | C19H15BF4 | 330.133 g/mol

It is applied in the abstraction of the-H atom of alcohols and their O-substituted derivatives results in oxidation to the ketones and in the oxidation of TMS ethers to ketones, secondary alcohols to ketones, and secondary diols to ?-hydroxy ketones. It is applied as a reactant involved in the synthesis of substituted dihydroazulene photo switches, carbene-based Lewis pairs for hydrogen activation, 1,3-Dipolar cycloaddition reactions for preparation of α-amino-β-hydroxy esters, oxidation of allenic compounds, Mukaiyama aldol addition reactions and ionic hydrogenation as a counteranion and ligand source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the abstraction of the ɑ-H atom of alcohols and their O-substituted derivatives results in oxidation to the ketones and in the oxidation of TMS ethers to ketones, secondary alcohols to ketones, and secondary diols to ɑ-hydroxy ketones. It is applied as a reactant involved in the synthesis of substituted dihydroazulene photo switches, carbene-based Lewis pairs for hydrogen activation, 1,3-Dipolar cycloaddition reactions for preparation of α-amino-β-hydroxy esters, oxidation of allenic compounds, Mukaiyama aldol addition reactions and ionic hydrogenation as a counteranion and ligand source.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Moisture sensitive. Store away from bases, water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 341-02-6
Molecular Formula C19H15BF4
Molecular Weight (g/mol) 330.133
MDL Number MFCD00013120
InChI Key VQXBOEYKSVVPSP-UHFFFAOYSA-N
Synonym triphenylcarbenium tetrafluoroborate, trityl tetrafluoroborate, tritylium tetrafluoroborate, triphenylmethylium tetrafluoroborate, diphenylmethylbenzene tetrafluoroborate, trityl fluoroborate, triphenylmethyl fluoroborate, triphenylcarbonium tetrafluoroborate, methylium, triphenyl-, tetrafluoroborate 1-, methylium, triphenyl-, tetrafluoroborate 1-1:1
PubChem CID 2723955
IUPAC Name diphenylmethylbenzene;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point ∼207°C (decomposition)
Odor Odorless
Linear Formula (C6H5)3CBF4
Quantity 250 g
UN Number UN3261
Beilstein 5085649
Sensitivity Moisture sensitive
Solubility Information Insoluble in water. Soluble in methanol.
Formula Weight 330.13
Percent Purity 98%
Chemical Name or Material Triphenylcarbenium tetrafluoroborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.