missing translation for 'onlineSavingsMsg'
Learn More

Triphenylcarbenium hexafluorophosphate, 98%

Product Code. 11332036
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332036 10 g 10g
11342036 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332036 Supplier Thermo Scientific Alfa Aesar Supplier No. L03140.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 437-17-2 | C19H15F6P | 388.29 g/mol

Triphenylcarbenium hexafluorophosphate is a reagent involved in Si-H and Si-Si bond activation at Pt of neutral and cationic silyl Pt bisphosphine complexes. It is used in investigations of interactions with chromium acetylacetonate nitride mononuclear complexes. It is used in double ring-opening polyaddition of spiro orthoesters. It is used as reagent in Synthesis of push-pull tropylium-fused aminoporphyrazine, Preparation of cycloheptatrienyl and tropylium calix[4]arenes, Synthesis of palladacycles containing chiral rhenium half-sandwich complex for use as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylcarbenium hexafluorophosphate is a reagent involved in Si-H and Si-Si bond activation at Pt of neutral and cationic silyl Pt bisphosphine complexes. It is used in investigations of interactions with chromium acetylacetonate nitride mononuclear complexes. It is used in double ring-opening polyaddition of spiro orthoesters. It is used as reagent in Synthesis of ″push-pull″ tropylium-fused aminoporphyrazine, Preparation of cycloheptatrienyl and tropylium calix[4]arenes, Synthesis of palladacycles containing chiral rhenium half-sandwich complex for use as a catalyst.

Solubility
Soluble in water. (27 g/L) at 25°C.

Notes
Moisture Sensitive. Store at 4°C. Store away from oxidizing agents and strong bases. Incompatible with heat and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437-17-2
Molecular Formula C19H15F6P
Molecular Weight (g/mol) 388.29
MDL Number MFCD00013121
InChI Key IBTFOFOFRZKIJU-UHFFFAOYSA-N
Synonym triphenylcarbenium hexafluorophosphate, trityl hexafluorophosphate, tritylium hexafluorophosphate, triphenylmethyl hexafluorophosphate, triphenylcarbonium hexafluorophosphate, tritylium, hexafluorophosphate 1-, triphenyl-methyliuhexafluorophosphate 1-, ph3cpf6, methylium, triphenyl-, hexafluorophosphate 1-, methylium, hexafluorophosphate 1-
PubChem CID 2723954
IUPAC Name diphenylmethylbenzene;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∽160°C (decomposition)
Linear Formula (C6H5)3CPF6
Quantity 10 g
UN Number UN3261
Beilstein 4344297
Sensitivity Moisture sensitive
Solubility Information Soluble in water. (27g/L) at 25°C.
Formula Weight 388.29
Percent Purity 98%
Chemical Name or Material Triphenylcarbenium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.