missing translation for 'onlineSavingsMsg'
Learn More

Triphenylamine, TRC

Product Code. 30453624
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453624 1 g 1g
30449912 10 g 10g
30453258 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453624 Supplier Toronto Research Chemicals Supplier No. TRCT8959951G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-34-9
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.32
Synonym Benzenamine, N,N-diphenyl- (9CI, ACI), N,N-Diphenylbenzenamine (ACI), Triphenylamine (8CI), N,N-Diphenylaniline, NSC 66458
IUPAC Name N,N-diphenylaniline
SMILES c1ccc(cc1)N(c2ccccc2)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H15N/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H
Formula Weight 245.1204
Chemical Name or Material Triphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.