missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl Phosphine, Pure, Fisher Chemical

Product Code. 10060450 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10060450 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10060450 Supplier Fisher Chemical Supplier No. T/3652/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 603-35-0
Melting Point 78.5°C
Color White
Boiling Point 377°C
Packaging Plastic powder jar
Molecular Formula C18H15P
MDL Number MFCD00003043 MFCD20489348
Quantity 100 g
Synonym triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin
InChI Key RIOQSEWOXXDEQQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name triphenylphosphane
Molecular Weight (g/mol) 262.29
PubChem CID 11776
Formula Weight 262.28g/mol
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triphenylphosphine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.