missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphate, TRC

Product Code. 30451632
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451632 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451632 Supplier Toronto Research Chemicals Supplier No. TRCT80899050G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triphenylphosphate
CAS 115-86-6
Purity Grade Notes HPLC
Molecular Formula C18 H15 O4 P
Quantity 50 g
Synonym Phosphoric acid','triphenyl ester, Celluflex TPP, DHPF 005, Disflamoll TP, NSC 57868, Phenyl phosphate ((PhO)3PO), Phoscon FR 903N, Phosflex TPP, Reofos TPP, S 4, S 4 (phosphate), Sumilizer TPP, TFF, TP, TPP, TPPA, TTP, TTP (triphenyl phosphate), Triphenol phosphate, Triphenoxyphosphine oxide, Triphenyl phosphate, WSFR-TPP, Wako TPP
InChI Formula InChI=1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H
SMILES O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3
IUPAC Name triphenyl phosphate
Molecular Weight (g/mol) 326.28
Formula Weight 326.0708
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.